Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tgif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291124.1 | NP_001278053.1 | 4 | 925 | ||
NM_173396.3 | NP_775572.1 | 3 | 1042 | |||
GenBank mRNA | AK017884.1 | - | 3 | 904 | ||
AK029490.1 | - | 3 | 972 | |||
AK157623.1 | - | 4 | 833 | |||
AK186967.1 | - | 1 | 51 | |||
BC138705.1 | - | 3 | 955 | 8861727 | ||
BC138706.1 | - | 3 | 957 | 8861728 | ||
BC158022.1 | - | 2 | 804 | 9007328 | ||
CF731285.1 | - | 3 | 543 |