Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mrpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175001.3 | NP_778166.2 | 7 | 613 | ||
GenBank mRNA | AK077615.1 | - | 7 | 607 | ||
AK086913.1 | - | 7 | 592 | |||
AK164417.1 | - | 7 | 592 | |||
BC061191.1 | - | 7 | 603 | 6703228 |