Search
Search

Dnali1| Gene Symbol: Dnali1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175223.4 | NP_780432.1 | 6 | 1507 | ||
| GenBank mRNA | AK077109.1 | - | 6 | 1508 | ||
| BC118619.1 | - | 6 | 1485 | 40107304 | ||
| BC122552.1 | - | 6 | 1482 | 40107303 | ||