Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Inpp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127363.1 | NP_001120835.1 | 6 | 688 | ||
NM_183144.3 | NP_898967.2 | 6 | 688 | |||
GenBank mRNA | AK083449.1 | - | 7 | 687 | ||
AK157558.1 | - | 6 | 551 | |||
AK171360.1 | - | 6 | 688 | |||
AK171880.1 | - | 6 | 664 | |||
BC056341.1 | - | 6 | 688 | 5703345 |