Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023622.4 | NP_076111.2 | 8 | 1071 | ||
GenBank mRNA | AF237669.1 | - | 8 | 1030 | ||
AF239661.1 | - | 8 | 1072 | |||
AK004876.1 | - | 7 | 804 | |||
AK053550.1 | - | 8 | 1058 | |||
AK131140.1 | - | 577 | ||||
AK134654.1 | - | 1053 | ||||
AK165023.1 | - | 8 | 1059 | |||
AK165746.1 | - | 8 | 1065 |