Search
Search

Kank3| Gene Symbol: Kank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030697.2 | NP_109622.1 | 9 | 2294 | ||
| GenBank mRNA | AK002623.1 | - | 9 | 2295 | ||
| AK132011.1 | - | 9 | 2316 | |||
| BC037741.1 | - | 4 | 428 | |||
| BC061055.1 | - | 4 | 440 | |||
| BC115933.1 | - | 3 | 398 | |||
| KJ650203.1 | - | 8 | 2225 | |||
| KJ663713.1 | - | 8 | 2066 | |||
| KJ663714.1 | - | 9 | 2087 | |||