Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INPP5D SIRNA Assays ›
          Gene Symbol:
          INPP5D
          Gene Name:
          inositol polyphosphate-5-phosphatase D
          Gene Aliases:
          SHIP, SHIP-1, SHIP1, SIP-145, hp51CN, p150Ship
          Chromosome Location:
          Chr.2: 233059967 - 233207903 on Build GRCh38
          UniGene ID:
          Hs.262886
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 16502

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: INPP5D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017915.2 NP_001017915.1 5 1066
          NM_005541.4 NP_005532.2 5 1063
          XM_017004004.1 XP_016859493.1 5 689
          GenBank mRNA AK298651.1 - 5 695
          AK309644.1 - 5 688
          BC099920.1 - 5 674 6304992
          BC113580.1 - 5 611 8322632
          BC113582.1 - 5 611 8322634
          U50041.1 - 5 675
          U53470.1 - 5 684
          U57650.1 - 5 1063
          U84400.1 - 5 635
          X98429.1 - 5 691

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylinositol biosynthetic process
          phosphate-containing compound metabolic process
          apoptotic process
          signal transduction
          determination of adult lifespan
          negative regulation of signal transduction
          immunoglobulin mediated immune response
          negative regulation of B cell proliferation
          intracellular signal transduction
          positive regulation of apoptotic process
          inositol phosphate metabolic process
          negative regulation of interleukin-6 biosynthetic process
          positive regulation of B cell differentiation
          positive regulation of erythrocyte differentiation
          negative regulation of monocyte differentiation
          negative regulation of neutrophil differentiation
          negative regulation of osteoclast differentiation
          negative regulation of bone resorption
          phosphatidylinositol dephosphorylation
          negative regulation of immune response
          T cell receptor signaling pathway
          leukocyte migration
          inositol-polyphosphate 5-phosphatase activity
          protein binding
          phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase activity
          SH3 domain binding
          phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
          PTB domain binding
          inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0