Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD44 SIRNA Assays ›
          Gene Symbol:
          Cd44
          Gene Name:
          CD44 antigen
          Gene Aliases:
          AU023126, AW121933, AW146109, HERMES, Ly-24, Pgp-1
          Chromosome Location:
          Chr.2: 102811141 - 102901669 on Build GRCm38
          UniGene ID:
          Mm.423621
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 165813

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cd44
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039150.1 NP_001034239.1 2 524
          NM_001039151.1 NP_001034240.1 2 524
          NM_001177785.1 NP_001171256.1 2 524
          NM_001177786.1 NP_001171257.1 2 524
          NM_001177787.1 NP_001171258.1 2 524
          NM_009851.2 NP_033981.2 2 524
          GenBank mRNA AJ251594.1 - 2 311
          AK045226.1 - 2 370
          AK087472.1 - 2 318
          AK144125.1 - 2 327
          AK152099.1 - 2 320
          AK154411.1 - 2 315
          AK162720.1 - 2 320
          AK166320.1 - 2 309
          BC005676.1 - 2 303
          BC051388.1 - 2 354 4910789
          CT010324.1 - 2 205
          J05163.1 - 2 186
          M27129.1 - 2 231
          M27130.1 - 2 237
          M30655.1 - 2 305
          X66081.1 - 2 208
          X66082.1 - 2 208
          X66083.1 - 2 208
          X66084.1 - 2 208

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          branching involved in ureteric bud morphogenesis
          wound healing involved in inflammatory response
          positive regulation of adaptive immune response
          negative regulation of mature B cell apoptotic process
          cell adhesion
          positive regulation of gene expression
          Wnt signaling pathway
          cell migration
          hyaluronan catabolic process
          neuron projection development
          positive regulation of neutrophil apoptotic process
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of heterotypic cell-cell adhesion
          macrophage fusion
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          cellular response to fibroblast growth factor stimulus
          positive regulation of peptidyl-tyrosine phosphorylation
          branching involved in prostate gland morphogenesis
          positive regulation of ERK1 and ERK2 cascade
          monocyte aggregation
          positive regulation of monocyte aggregation
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          hyalurononglucosaminidase activity
          cytokine receptor activity
          epidermal growth factor receptor binding
          protein binding
          hyaluronic acid binding
          protein kinase binding
          cytokine binding
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline