Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ryk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042607.1 | NP_001036072.1 | 3 | 531 | ||
NM_001284258.1 | NP_001271187.1 | 2 | 409 | |||
NM_013649.3 | NP_038677.3 | 3 | 531 | |||
GenBank mRNA | AK144355.1 | - | 2 | 349 | ||
BC032275.1 | - | 3 | 265 | 5370100 | ||
L02210.1 | - | 3 | 506 | |||
L21707.1 | - | 3 | 541 | |||
L38394.1 | - | 3 | 454 |