Search Thermo Fisher Scientific
Gene Symbol: Atad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026487.3 | NP_080763.2 | 7 | 814 | ||
GenBank mRNA | AK007147.1 | - | 3 | 271 | ||
AK009419.1 | - | 3 | 402 | |||
AK014967.1 | - | 7 | 831 | |||
AK030719.1 | - | 7 | 834 | |||
AK033639.1 | - | 7 | 838 | |||
AK044925.1 | - | 7 | 814 | |||
AK150469.1 | - | 7 | 834 | |||
AK152059.1 | - | 7 | 834 | |||
AK165953.1 | - | 7 | 849 | |||
BC029085.1 | - | 7 | 814 | 3593451 | ||
BC043051.1 | - | 7 | 799 | 5708377 |