Search
Search

Arhgap17| Gene Symbol: Arhgap17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122640.1 | NP_001116112.1 | 15, 16 | 1452 | ||
| NM_001122641.1 | NP_001116113.1 | 15, 16 | 1452 | |||
| NM_001122642.1 | NP_001116114.1 | 15, 16 | 1452 | |||
| NM_001122643.1 | NP_001116115.1 | 15, 16 | 1452 | |||
| NM_144529.2 | NP_653112.2 | 15, 16 | 1452 | |||
| GenBank mRNA | AB060553.1 | - | 15, 16 | 1450 | ||
| AB060554.1 | - | 15, 16 | 1450 | |||
| AK036468.1 | - | 15, 16 | 1389 | |||
| AK036617.1 | - | 15, 16 | 1389 | |||
| AK046411.1 | - | 14, 15 | 1359 | |||
| AK147010.1 | - | 15, 16 | 1423 | |||
| AK149986.1 | - | 15, 16 | 1424 | |||
| AK152266.1 | - | 15, 16 | 1389 | |||
| AK152301.1 | - | 15, 16 | 1416 | |||
| AK161636.1 | - | 15 | 2379 | |||
| AK179915.1 | - | 1, 2 | 66 | |||
| BC003259.1 | - | 15, 16 | 1400 | 3500822 | ||