Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tas2r117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207021.1 | NP_996904.1 | 1 | 559 | ||
GenBank mRNA | BC132206.1 | - | 1 | 642 | 40130483 | |
BC132208.1 | - | 1 | 642 | 40130485 |