Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ephb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010143.1 | NP_034273.1 | 3 | 596 | ||
GenBank mRNA | AK144414.1 | - | 3 | 612 | ||
AK158043.1 | - | 3 | 1657 | |||
BC014822.1 | - | 3 | 597 | 3673003 | ||
BC053085.1 | - | 3 | 596 | 5708827 | ||
U11493.1 | - | 1 | 56 | |||
Z49086.1 | - | 3 | 454 |