Search
Search

Ecsit| Gene Symbol: Ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253897.1 | NP_001240826.1 | 9 | 1388 | ||
| NM_001253898.1 | NP_001240827.1 | 8 | 1210 | |||
| NM_012029.2 | NP_036159.1 | 9 | 1396 | |||
| GenBank mRNA | AF182510.1 | - | 8 | 1409 | ||
| AK007564.1 | - | 9 | 1362 | |||
| AK012835.1 | - | 9 | 1361 | |||
| BC004583.1 | - | 8 | 1197 | 3587327 | ||
| BC016424.1 | - | 9 | 1336 | 4924678 | ||