Search Thermo Fisher Scientific
Gene Symbol: Myo1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164573.1 | NP_001158045.1 | 27 | 2756 | ||
GenBank mRNA | AK014505.1 | - | 6 | 686 | ||
BC144867.1 | - | 6 | 661 | 9053415 | ||
BC144868.1 | - | 6 | 662 | |||
BC144869.1 | - | 6 | 589 | |||
BC144870.1 | - | 6 | 637 | 9053418 | ||
BC144872.1 | - | 5 | 605 |