Search
Search

Dnm1l| Gene Symbol: Dnm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025947.2 | NP_001021118.1 | 11 | 1496 | ||
| NM_001276340.1 | NP_001263269.1 | 11 | 1514 | |||
| NM_001276341.1 | NP_001263270.1 | 10 | 1258 | |||
| NM_152816.3 | NP_690029.2 | 12 | 1535 | |||
| GenBank mRNA | AB079133.1 | - | 11 | 1348 | ||
| AK018195.1 | - | 12 | 1475 | |||
| AK051443.1 | - | 11 | 1425 | |||
| AK080871.1 | - | 11 | 1416 | |||
| AK154123.1 | - | 11 | 1265 | |||
| AK190489.1 | - | 1 | 76 | |||
| BC040777.1 | - | 10 | 1228 | 4948368 | ||
| BC079635.1 | - | 11 | 1345 | 6400313 | ||
| BC085158.1 | - | 12 | 1440 | |||
| BQ550714.1 | - | 3 | 432 | |||
| CD559921.1 | - | 3 | 358 | |||
| CK391623.1 | - | 3 | 283 | |||
| DV652251.1 | - | 4 | 522 | |||