Search
Search

Adam32| Gene Symbol: Adam32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293693.1 | NP_001280622.1 | 9 | 806 | ||
| NM_153397.2 | NP_700446.2 | 9 | 806 | |||
| GenBank mRNA | AF513715.1 | - | 9 | 789 | ||
| AK005759.1 | - | 9 | 797 | |||
| BC060983.1 | - | 9 | 803 | 6772485 | ||