Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Skint7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142775.1 | NP_001136247.1 | 3 | 308 | ||
NM_177818.3 | NP_808486.2 | 3 | 308 | |||
GenBank mRNA | AK048392.1 | - | 3 | 203 | ||
AK132639.1 | - | 3 | 313 | |||
BB614300.1 | - | 3 | 308 | |||
EF494899.1 | - | 146 | ||||
EF494900.1 | - | 146 | ||||
EF494901.1 | - | 146 | ||||
EF494902.1 | - | 146 | ||||
EU099306.1 | - | 146 |
Gene Symbol: Skint8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100466.1 | NP_001093936.2 | 3 | 156 | ||
GenBank mRNA | AK076260.1 | - | 2 | 2210 | ||
EF494903.1 | - | 3 | 156 | |||
EU099307.1 | - | 3 | 156 | |||
EU099308.1 | - | 3 | 156 |