Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pigl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039536.2 | NP_001034625.1 | 1, 2 | 243 | ||
GenBank mRNA | AK128932.1 | - | 1, 2 | 243 | ||
AK158149.1 | - | 1, 2 | 244 |