Search
Search

Rab21| Gene Symbol: Rab21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024454.1 | NP_077774.1 | 5 | 666 | ||
| GenBank mRNA | AB232615.1 | - | 5 | 421 | ||
| AK035980.1 | - | 4 | 235 | |||
| AK172891.1 | - | 5 | 666 | |||
| BC055042.1 | - | 5 | 666 | 6490069 | ||