Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zgpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048148.1 | NP_001041613.1 | 2 | 887 | ||
NM_144894.3 | NP_659143.1 | 3 | 1111 | |||
GenBank mRNA | AK041491.1 | - | 3 | 1115 | ||
AK049764.1 | - | 2 | 886 | |||
AK052236.1 | - | 2 | 785 | |||
AK159801.1 | - | 2 | 906 | |||
BC021513.1 | - | 2 | 885 | 5351756 | ||
BC027218.1 | - | 2 | 794 | 3158198 |