Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145478.2 | NP_663453.1 | 6 | 2314 | ||
GenBank mRNA | AK079623.1 | - | 6 | 2039 | ||
AK149478.1 | - | 6 | 2034 | |||
AK159399.1 | - | 6 | 2032 | |||
AK159600.1 | - | 6 | 2032 | |||
AK159670.1 | - | 6 | 2009 | |||
AK168014.1 | - | 6 | 2323 | |||
AK188587.1 | - | 1 | 194 | |||
AK189962.1 | - | 1 | 194 | |||
AK197499.1 | - | 1 | 194 | |||
AK209684.1 | - | 1 | 193 | |||
AK210582.1 | - | 1 | 194 | |||
AK213303.1 | - | 1 | 169 | |||
AK214252.1 | - | 1 | 194 | |||
AY026239.1 | - | 6 | 2057 | |||
BC017621.1 | - | 6 | 2314 | 4924687 | ||
BC026639.1 | - | 6 | 2003 | 4985551 |