Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282055.1 | NP_001268984.1 | 15 | 2112 | ||
GenBank mRNA | AK088950.1 | - | 14 | 3702 | ||
AK129220.1 | - | 14 | 3703 | |||
AK134073.1 | - | 3 | 1358 | |||
BC026492.1 | - | 8 | 1057 | 4505095 | ||
BC092240.1 | - | 14 | 2023 | 30662573 |