Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABI2 SIRNA Assays ›
          Gene Symbol:
          ABI2
          Gene Name:
          abl-interactor 2
          Gene Aliases:
          ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
          Chromosome Location:
          Chr.2: 203327874 - 203432173 on Build GRCh38
          UniGene ID:
          Hs.471156
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 17004

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABI2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282925.1 NP_001269854.1 11 1707
          NM_001282926.1 NP_001269855.1 10 1689
          NM_001282927.1 NP_001269856.1 9 1182
          NM_001282932.1 NP_001269861.1 10 1703
          NM_005759.5 NP_005750.4 10 1593
          XM_005246217.2 XP_005246274.1 12 2159
          XM_005246218.2 XP_005246275.1 11 2141
          XM_005246221.2 XP_005246278.1 10 1994
          XM_005246225.2 XP_005246282.1 9 1907
          XM_005246226.2 XP_005246283.1 9 1871
          XM_006712174.2 XP_006712237.1 12 2159
          XM_006712177.2 XP_006712240.1 13 2235
          XM_006712179.2 XP_006712242.1 11 2012
          XM_006712180.2 XP_006712243.1 11 1976
          XM_006712181.2 XP_006712244.1 10 1889
          XM_006712182.2 XP_006712245.1 10 1886
          XM_006712185.1 XP_006712248.1 10 1947
          XM_006712186.2 XP_006712249.1 8 1724
          XM_011510472.2 XP_011508774.1 11 2009
          XM_011510473.2 XP_011508775.1 11 1458
          XM_017003118.1 XP_016858607.1 12 2156
          XM_017003119.1 XP_016858608.1 12 2153
          XM_017003120.1 XP_016858609.1 12 2156
          XM_017003121.1 XP_016858610.1 11 2069
          XM_017003122.1 XP_016858611.1 11 2006
          XM_017003123.1 XP_016858612.1 10 1991
          XM_017003124.1 XP_016858613.1 10 1958
          XM_017003125.1 XP_016858614.1 10 1955
          XM_017003126.1 XP_016858615.1 10 1952
          XM_017003127.1 XP_016858616.1 10 1955
          XM_017003128.1 XP_016858617.1 12 2148
          XM_017003129.1 XP_016858618.1 10 1925
          XM_017003130.1 XP_016858619.1 10 1922
          XM_017003131.1 XP_016858620.1 9 1904
          XM_017003132.1 XP_016858621.1 10 1889
          XM_017003133.1 XP_016858622.1 12 2088
          XM_017003134.1 XP_016858623.1 9 1871
          XM_017003135.1 XP_016858624.1 9 1868
          XM_017003136.1 XP_016858625.1 9 1868
          XM_017003137.1 XP_016858626.1 12 2049
          XM_017003138.1 XP_016858627.1 10 1826
          XM_017003139.1 XP_016858628.1 11 2034
          XM_017003140.1 XP_016858629.1 11 2031
          XM_017003141.1 XP_016858630.1 9 1811
          XM_017003142.1 XP_016858631.1 9 1808
          XM_017003143.1 XP_016858632.1 11 1998
          XM_017003144.1 XP_016858633.1 9 1742
          XM_017003145.1 XP_016858634.1 8 1721
          XM_017003146.1 XP_016858635.1 8 1724
          GenBank mRNA AF260261.1 - 11 1431
          AK294429.1 - 5 759
          AK297537.1 - 6 648
          AK299092.1 - 9 925
          AK299672.1 - 10 1084
          AK299679.1 - 5 625
          AK299688.1 - 7 885
          AK299750.1 - 5 531
          AK299824.1 - 10 1533
          AK302853.1 - 10 1629
          AK316343.1 - 5 585
          BC001439.2 - 10 1342 3138341
          BT009920.1 - 10 1317
          U23435.1 - 9 1143
          U31089.1 - 9 1062
          X95632.1 - 10 1417

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          cytoskeleton organization
          actin polymerization or depolymerization
          viral process
          cell migration
          Rac protein signal transduction
          peptidyl-tyrosine phosphorylation
          positive regulation of Arp2/3 complex-mediated actin nucleation
          DNA binding
          protein binding
          cytoskeletal adaptor activity
          SH3 domain binding
          kinase binding
          ubiquitin protein ligase binding
          protein complex binding
          Rac GTPase binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5hctl:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0