Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bud13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146000.2 | NP_666112.1 | 5 | 1277 | ||
GenBank mRNA | AK037134.1 | - | 5 | 1274 | ||
AK038126.1 | - | 5 | 1282 | |||
AK051047.1 | - | 5 | 1282 | |||
AK077744.1 | - | 5 | 1281 | |||
BC025490.1 | - | 5 | 1278 | 5252666 |