Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CTDSPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008392.1 | NP_001008393.1 | 8 | 3400 | ||
NM_005808.2 | NP_005799.2 | 7 | 3367 | |||
XM_017005519.1 | XP_016861008.1 | 9 | 2465 | |||
XM_017005520.1 | XP_016861009.1 | 9 | 3783 | |||
GenBank mRNA | D88153.1 | - | 8 | 3713 |