Search
Search

NET1| Gene Symbol: NET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047160.2 | NP_001040625.1 | 7 | 789 | ||
| NM_005863.4 | NP_005854.2 | 5 | 700 | |||
| NR_073040.1 | 5 | 733 | ||||
| GenBank mRNA | AB209852.1 | - | 7 | 735 | ||
| AJ010046.1 | - | 5 | 633 | |||
| AK024919.1 | - | 5 | 640 | |||
| AK299335.1 | - | 7 | 745 | |||
| AK302687.1 | - | 7 | 571 | |||
| AK304028.1 | - | 5 | 675 | |||
| AK315566.1 | - | 5 | 642 | |||
| BC010285.1 | - | 5 | 631 | 3048980 | ||
| BC053553.1 | - | 7 | 741 | 6137683 | ||
| BX537509.1 | - | 2 | 2913 | |||
| U02081.1 | - | 3 | 221 | |||