Search
Search

Kansl3| Gene Symbol: Kansl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172652.3 | NP_766240.1 | 2 | 306 | ||
| GenBank mRNA | AK014565.1 | - | 2 | 264 | ||
| AK030079.1 | - | 2 | 307 | |||
| AK129329.1 | - | 2 | 159 | |||
| BC132293.1 | - | 1 | 209 | 40130570 | ||
| BC144763.1 | - | 1 | 209 | |||