Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tapt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173764.3 | NP_776125.2 | 14 | 1607 | ||
GenBank mRNA | AK030002.1 | - | 9 | 849 | ||
BC096038.1 | - | 11 | 1086 | |||
BC110670.1 | - | 13 | 1328 |