Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYP26B1 SIRNA Assays ›
          Gene Symbol:
          Cyp26b1
          Gene Name:
          cytochrome P450, family 26, subfamily b, polypeptide 1
          Gene Aliases:
          CP26, P450RAI-2
          Chromosome Location:
          Chr.6: 84571414 - 84593908 on Build GRCm38
          UniGene ID:
          Mm.255246
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 171397

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cyp26b1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177713.1 NP_001171184.1 4 874
          NM_175475.3 NP_780684.1 4 925
          GenBank mRNA AK028272.1 - 2 409
          AK137124.1 - 3 1200
          AK166211.1 - 3 463
          AY134662.1 - 4 861
          BC059246.1 - 4 874 6400154

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate determination
          establishment of T cell polarity
          inflammatory response
          male meiosis
          spermatogenesis
          proximal/distal pattern formation
          positive regulation of gene expression
          embryonic limb morphogenesis
          retinoic acid catabolic process
          retinoic acid metabolic process
          tongue morphogenesis
          regulation of T cell differentiation
          retinoic acid receptor signaling pathway
          regulation of retinoic acid receptor signaling pathway
          negative regulation of retinoic acid receptor signaling pathway
          oxidation-reduction process
          bone morphogenesis
          establishment of skin barrier
          cornification
          positive regulation of tongue muscle cell differentiation
          retinoic acid binding
          monooxygenase activity
          iron ion binding
          retinoic acid 4-hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          fluorene oxygenase activity
          mono-butyltin dioxygenase activity
          tri-n-butyltin dioxygenase activity
          di-n-butyltin dioxygenase activity
          methylsilanetriol hydroxylase activity
          methyl tertiary butyl ether 3-monooxygenase activity
          4-nitrocatechol 4-monooxygenase activity
          4-chlorophenoxyacetate monooxygenase activity
          tert-butanol 2-monooxygenase activity
          alpha-pinene monooxygenase activity
          dimethylsilanediol hydroxylase activity
          ammonia monooxygenase activity
          hydroxymethylsilanetriol oxidase activity
          2-hydroxyisobutyrate 3-monooxygenase activity
          alpha-pinene dehydrogenase activity
          heme binding
          bisphenol A hydroxylase B activity
          2,2-bis(4-hydroxyphenyl)-1-propanol hydroxylase activity
          9-fluorenone-3,4-dioxygenase activity
          anthracene 9,10-dioxygenase activity
          2-(methylthio)benzothiazole monooxygenase activity
          2-hydroxybenzothiazole monooxygenase activity
          benzothiazole monooxygenase activity
          2,6-dihydroxybenzothiazole monooxygenase activity
          pinacolone 5-monooxygenase activity
          thioacetamide S-oxygenase activity
          thioacetamide S-oxide S-oxygenase activity
          endosulfan monooxygenase I activity
          N-nitrodimethylamine hydroxylase activity
          4-(1-ethyl-1,4-dimethyl-pentyl)phenol monoxygenase activity
          endosulfan ether monooxygenase activity
          pyrene 4,5-monooxygenase activity
          pyrene 1,2-monooxygenase activity
          1-hydroxypyrene 6,7-monooxygenase activity
          1-hydroxypyrene 7,8-monooxygenase activity
          phenylboronic acid monooxygenase activity
          spheroidene monooxygenase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0