Search
Search

Ldoc1l| Gene Symbol: Ldoc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177630.3 | NP_808298.2 | 1 | 1407 | ||
| GenBank mRNA | AK083881.1 | - | 1 | 1369 | ||
| BC035491.1 | - | 1 | 463 | |||
| BC058638.1 | - | 1 | 1076 | 6418256 | ||
| BC094559.1 | - | 1 | 1407 | 6826763 | ||