Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lppr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177664.5 | NP_808332.3 | 3 | 1102 | ||
GenBank mRNA | AF541279.1 | - | 3 | 1103 | ||
AK046782.1 | - | 3 | 1103 | |||
AK079635.1 | - | 3 | 436 | |||
AK129149.1 | - | 3 | 698 | |||
AY266266.1 | - | 3 | 1103 | |||
AY266267.1 | - | 3 | 1103 | |||
BC082323.1 | - | 3 | 632 | |||
BC137701.1 | - | 3 | 697 | 8860720 | ||
BC137702.1 | - | 3 | 698 | 8860721 |