Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Alkbh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198027.2 | NP_932144.2 | 7 | 869 | ||
GenBank mRNA | AK082058.1 | - | 6 | 1037 | ||
BC029805.1 | - | 7 | 816 | 4952089 | ||
BC080310.1 | - | 5 | 992 |