Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198168.3 | NP_937811.1 | 12, 13 | 1838 | ||
GenBank mRNA | AK034167.1 | - | 12, 13 | 1857 | ||
AK155872.1 | - | 12, 13 | 1831 | |||
AK171041.1 | - | 12, 13 | 1809 | |||
AK171178.1 | - | 12, 13 | 1839 | |||
AK171430.1 | - | 12, 13 | 1846 | |||
BC026670.1 | - | 8, 9 | 694 | |||
BC058977.1 | - | 12, 13 | 1629 | 5687054 |