Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rbm11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198302.2 | NP_938044.1 | 2 | 239 | ||
GenBank mRNA | AK039297.1 | - | 2 | 181 | ||
BC050779.1 | - | 2 | 286 | 6771154 |