Search
Search

Pde6c| Gene Symbol: Pde6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170959.1 | NP_001164430.1 | 2, 3 | 806 | ||
| NM_033614.2 | NP_291092.1 | 2, 3 | 806 | |||
| GenBank mRNA | AF411063.1 | - | 2, 3 | 641 | ||
| BC027050.1 | - | 2, 3 | 806 | 5358611 | ||