Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mtmr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133215.1 | NP_573478.1 | 14 | 1644 | ||
GenBank mRNA | AF262986.1 | - | 14 | 1644 | ||
AK146641.1 | - | 14 | 1661 | |||
AK186095.1 | - | 2 | 213 |