Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DICER1 SIRNA Assays ›
          Gene Symbol:
          Dicer1
          Gene Name:
          dicer 1, ribonuclease type III
          Gene Aliases:
          1110006F08Rik, D12Ertd7e, mKIAA0928
          Chromosome Location:
          Chr.12: 104687740 - 104751952 on Build GRCm38
          UniGene ID:
          Mm.21135
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 173425

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dicer1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_148948.2 NP_683750.2 4, 5 695
          GenBank mRNA AB081470.1 - 3, 4 493
          AF430845.1 - 4, 5 703
          AK035253.1 - 1, 2 40
          BC019805.1 - 4, 5 694
          BC061198.1 - 5, 6 643

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_030621, NM_001271282, NM_001195573, NM_177438, AB028449, and others.
          This siRNA also targets the following transcripts in Rat: XM_008764904.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          meiotic spindle organization
          angiogenesis
          regulation of protein phosphorylation
          hair follicle development
          apoptotic DNA fragmentation
          RNA processing
          cell proliferation
          regulation of Notch signaling pathway
          post-embryonic development
          zygote asymmetric cell division
          regulation of gene expression
          negative regulation of Schwann cell proliferation
          positive regulation of gene expression
          regulation of muscle cell apoptotic process
          positive regulation of Schwann cell differentiation
          myoblast differentiation involved in skeletal muscle regeneration
          rRNA catabolic process
          stem cell population maintenance
          spinal cord motor neuron differentiation
          nerve development
          olfactory bulb interneuron differentiation
          cerebral cortex development
          lung development
          embryonic limb morphogenesis
          production of siRNA involved in RNA interference
          targeting of mRNA for destruction involved in RNA interference
          gene silencing by RNA
          pre-miRNA processing
          hair follicle morphogenesis
          regulation of myelination
          positive regulation of myelination
          peripheral nervous system myelin formation
          conversion of ds siRNA to ss siRNA involved in RNA interference
          siRNA loading onto RISC involved in RNA interference
          embryonic hindlimb morphogenesis
          production of miRNAs involved in gene silencing by miRNA
          multicellular organism growth
          miRNA loading onto RISC involved in gene silencing by miRNA
          conversion of ds siRNA to ss siRNA
          regulation of odontogenesis of dentin-containing tooth
          regulation of viral genome replication
          regulation of cell differentiation
          regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          mRNA stabilization
          spleen development
          digestive tract development
          reproductive structure development
          regulation of oligodendrocyte differentiation
          epidermis morphogenesis
          branching morphogenesis of an epithelial tube
          neuron projection morphogenesis
          anatomical structure development
          regulation of neurogenesis
          cartilage development
          spindle assembly
          regulation of RNA metabolic process
          defense response to virus
          regulation of cell cycle
          cardiac muscle cell development
          inner ear receptor cell development
          negative regulation of glial cell proliferation
          intestinal epithelial cell development
          cardiac neural crest cell development involved in outflow tract morphogenesis
          ganglion development
          regulation of enamel mineralization
          hair follicle cell proliferation
          RNA phosphodiester bond hydrolysis
          RNA phosphodiester bond hydrolysis, endonucleolytic
          regulation of miRNA metabolic process
          positive regulation of miRNA metabolic process
          nucleotide binding
          nucleic acid binding
          RNA binding
          double-stranded RNA binding
          helicase activity
          nuclease activity
          endonuclease activity
          endoribonuclease activity
          ribonuclease III activity
          deoxyribonuclease I activity
          protein binding
          ATP binding
          hydrolase activity
          endoribonuclease activity, producing 5'-phosphomonoesters
          protein domain specific binding
          siRNA binding
          miRNA binding
          metal ion binding
          pre-miRNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0