Search
Search

KLHL41| Gene Symbol: KLHL41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006063.2 | NP_006054.2 | 6 | 1834 | ||
| GenBank mRNA | AF056929.1 | - | 6 | 1804 | ||
| AF333387.1 | - | 6 | 1834 | |||
| AK316571.1 | - | 6 | 1831 | |||
| AY364252.1 | - | 6 | 1244 | |||
| BC006534.2 | - | 6 | 1812 | 2959774 | ||