Search
Search

Rars2| Gene Symbol: Rars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181406.3 | NP_852071.2 | 3, 4 | 254 | ||
| GenBank mRNA | AK075721.1 | - | 3, 4 | 239 | ||
| AK156182.1 | - | 3, 4 | 232 | |||
| AK166986.1 | - | 3, 4 | 770 | |||
| BC024878.1 | - | 3, 4 | 234 | 3989666 | ||
| BU961647.1 | - | 256 | ||||