Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Taf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025444.2 | NP_079720.1 | 4 | 1178 | ||
GenBank mRNA | AK007339.1 | - | 4 | 1179 | ||
AK076595.1 | - | 1178 | ||||
AK160171.1 | - | 4 | 1178 | |||
BC021447.1 | - | 4 | 1190 | 5050736 |