Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOL3 SIRNA Assays ›
Gene Symbol:
Nol3
Gene Name:
nucleolar protein 3 (apoptosis repressor with CARD domain)
Gene Aliases:
ARC, B430311C09Rik, MYC, NOP, Nop30
Chromosome Location:
Chr.8: 105275937 - 105281939 on Build GRCm38
UniGene ID:
Mm.475715
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
174806
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nol3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_030152.4
NP_084428.1
4
1441
GenBank mRNA
AK053846.1
-
4
1442
AK079536.1
-
4
1473
AK197583.1
-
1
128
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
blood vessel remodeling
response to ischemia
mRNA splice site selection
mRNA processing
regulation of gene expression
cardiac muscle cell apoptotic process
negative regulation of striated muscle cell apoptotic process
negative regulation of cardiac muscle cell apoptotic process
negative regulation of tumor necrosis factor-mediated signaling pathway
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
negative regulation of muscle atrophy
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
response to injury involved in regulation of muscle adaptation
regulation of NF-kappaB import into nucleus
regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
myoblast differentiation
smooth muscle cell proliferation
protein oligomerization
negative regulation of cytosolic calcium ion concentration
negative regulation of mitochondrial calcium ion concentration
negative regulation of necrotic cell death
cellular response to hypoxia
negative regulation of release of cytochrome c from mitochondria
intrinsic apoptotic signaling pathway
inhibition of cysteine-type endopeptidase activity
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of mitochondrial membrane permeability involved in apoptotic process
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
negative regulation of death-inducing signaling complex assembly
negative regulation of hydrogen peroxide-induced cell death
negative regulation of protein targeting to mitochondrion
negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
receptor binding
death receptor binding
calcium ion binding
protein binding
kinase binding
phosphatase binding
death effector domain binding
identical protein binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
metal ion binding
caspase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0