Search Thermo Fisher Scientific
Gene Symbol: Dbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031403.3 | NP_113580.2 | 8 | 1223 | ||
GenBank mRNA | AK050090.1 | - | 8 | 1224 | ||
AK090351.1 | - | 8 | 1199 | |||
AK139631.1 | - | 8 | 1207 | |||
BC006661.1 | - | 8 | 1218 | 3501390 |