Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Mapre3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133350.1 | NP_579928.1 | 2 | 245 | ||
GenBank mRNA | AF468005.1 | - | 2 | 245 | ||
AK177097.1 | - | 2 | 160 | |||
BC048833.1 | - | 2 | 247 | |||
BC057918.1 | - | 2 | 245 | 5134461 |