Search
Search

VAV3| Gene Symbol: VAV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079874.1 | NP_001073343.1 | 10 | 1628 | ||
| NM_006113.4 | NP_006104.4 | 27 | 3289 | |||
| XM_005270360.2 | XP_005270417.1 | 26 | 3101 | |||
| XM_017000053.1 | XP_016855542.1 | 25 | 3210 | |||
| XM_017000055.1 | XP_016855544.1 | 26 | 2959 | |||
| XM_017000056.1 | XP_016855545.1 | 16 | 2165 | |||
| GenBank mRNA | AF067817.1 | - | 27 | 3272 | ||
| AF118887.1 | - | 27 | 3281 | |||
| BC143969.1 | - | 28 | 3456 | 9052487 | ||