Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012518.3 | NP_001012536.2 | 17 | 2551 | ||
NM_001109686.2 | NP_001103156.1 | 16 | 2392 | |||
NM_001109687.2 | NP_001103157.1 | 16 | 2407 | |||
NM_172545.4 | NP_766133.2 | 17 | 2530 | |||
GenBank mRNA | AB205007.1 | - | 17 | 2557 | ||
AK049454.1 | - | 14 | 1801 | |||
AK052174.1 | - | 17 | 2514 | |||
AK082062.1 | - | 17 | 2541 | |||
BC056938.1 | - | 14 | 2108 | |||
BC089302.1 | - | 14 | 2260 |