Search
Search

Ahcyl2| Gene Symbol: Ahcyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171000.2 | NP_001164471.2 | 10, 11 | 1397 | ||
| NM_001171001.1 | NP_001164472.1 | 10, 11 | 1117 | |||
| NM_021414.6 | NP_067389.5 | 10, 11 | 1400 | |||
| GenBank mRNA | AK014539.1 | - | 10, 11 | 1348 | ||
| AK053527.1 | - | 10, 11 | 1081 | |||
| AK122382.1 | - | 8, 9 | 1103 | |||
| BC051504.1 | - | 30 | ||||
| BC059255.1 | - | 6, 7 | 572 | |||
| BC079660.1 | - | 10, 11 | 1310 | 6853550 | ||