Search
Search

Ahcyl2| Gene Symbol: Ahcyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171000.2 | NP_001164471.2 | 16, 17 | 1935 | ||
| NM_001171001.1 | NP_001164472.1 | 16, 17 | 1655 | |||
| NM_021414.6 | NP_067389.5 | 16, 17 | 1938 | |||
| GenBank mRNA | AK014539.1 | - | 17, 18 | 1889 | ||
| AK053527.1 | - | 16, 17 | 1619 | |||
| AK122382.1 | - | 14, 15 | 1641 | |||
| AK161892.1 | - | 5, 6 | 415 | |||
| BC037447.1 | - | 4, 5 | 382 | |||
| BC051504.1 | - | 6, 7 | 568 | |||
| BC059255.1 | - | 12, 13 | 1110 | |||
| BC079660.1 | - | 16, 17 | 1848 | 6853550 | ||