Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTGER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000956.3 | NP_000947.2 | 2 | 1138 | ||
GenBank mRNA | AY275471.1 | - | 2 | 887 | ||
BC093927.1 | - | 2 | 940 | 7939772 | ||
BC093929.1 | - | 2 | 940 | 7939774 | ||
U19487.1 | - | 2 | 1043 | |||
X83868.1 | - | 2 | 887 |
Set Membership: |
G-protein coupled receptor |