Search
Search

Ifih1| Gene Symbol: Ifih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164477.1 | NP_001157949.1 | 1 | 729 | ||
| NM_027835.3 | NP_082111.2 | 1 | 729 | |||
| GenBank mRNA | AF374384.1 | - | 1 | 638 | ||
| AK037057.1 | - | 1 | 627 | |||
| AK040519.1 | - | 1 | 686 | |||
| AK149802.1 | - | 1 | 636 | |||
| AK153018.1 | - | 1 | 636 | |||
| AK156450.1 | - | 1 | 637 | |||
| AK168646.1 | - | 1 | 730 | |||
| AY075132.1 | - | 1 | 404 | |||
| BC080200.1 | - | 1 | 634 | 3597815 | ||
| EF113547.1 | - | 1 | 418 | |||
| EF113548.1 | - | 1 | 418 | |||
| EU084750.1 | - | 1 | 418 | |||